4-Amino-2-fluoro-N-methylbenzamide 98%

Supplier: Ambeed
Warning

A197283-100G A197283-10G A197283-1G A197283-250MG A197283-25G A197283-5G
76825-440EA 264 USD
76825-440 76825-442 76825-444 76825-446 76825-448 76825-450
4-Amino-2-fluoro-N-methylbenzamide 98%
4-Amino-2-fluoro-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₉FN₂O
MDL Number: MFCD09909322
CAS Number: 915087-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR