1-(1H-Benzo[d][1,2,3]triazol-1-yl)octan-1-one 97%
Supplier: Ambeed
|
Warning
|
A196589-100G
A196589-1G
A196589-250MG
A196589-25G
A196589-500G
A196589-5G
76825-412EA
180.36
USD
76825-412
76825-414
76825-416
76825-418
76825-420
76825-422
1-(1H-Benzo[d][1,2,3]triazol-1-yl)octan-1-one 97%
1-(1H-Benzo[d][1,2,3]triazol-1-yl)octan-1-one
Precaución: for Research Use Only
Formula:
C₁₄H₁₉N₃O MW: 245.32 g/mol |
MDL Number:
MFCD00690106 CAS Number: 58068-80-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...