1-Boc-4-(6-nitro-3-pyridyl)piperazine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 4-(6-nitropyridin-3-yl)piperazine-1-carboxylate, 4-(6-Nitropyridin-3-yl)piperazine, N1-BOC protected
A196536-10G
A196536-1G
A196536-250MG
A196536-25G
A196536-5G
76825-402EA
30
USD
76825-402
76825-404
76825-406
76825-408
76825-410
1-Boc-4-(6-nitro-3-pyridyl)piperazine 97%
1-Boc-4-(6-nitro-3-pyridyl)piperazine
Precaución: for Research Use Only
Formula:
C₁₄H₂₀N₄O₄ MW: 308.33 g/mol |
MDL Number:
MFCD11849291 CAS Number: 571189-16-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...