5-(2-Fluorophenyl)pyrrole-3-carboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 5-(2-Fluorophenyl)-1H-pyrrole-3-carbaldehyde

A196437-10G A196437-1G A196437-250MG A196437-25G A196437-5G
76825-392EA 48.81 USD
76825-392 76825-394 76825-396 76825-398 76825-400
5-(2-Fluorophenyl)pyrrole-3-carboxaldehyde 98%
5-(2-Fluorophenyl)pyrrole-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₈FNO
MW: 189.19 g/mol
MDL Number: MFCD11875867
CAS Number: 881674-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR