1-[3,5-Bis(trifluoromethyl)phenyl]-3-[(1R,2R)-(-)-2-(dimethylamino)cyclohexyl]thiourea 96%

Supplier: Ambeed
Warning

Synonyms: 1-(3,5-Bis(trifluoromethyl)phenyl)-3-((1R,2R)-2-(dimethylamino)cyclohexyl)thiourea, (R,R-TUC)

A196369-10G A196369-1G A196369-250MG A196369-25G
76825-382EA 491 USD
76825-382 76825-384 76825-386 76825-388
1-[3,5-Bis(trifluoromethyl)phenyl]-3-[(1R,2R)-(-)-2-(dimethylamino)cyclohexyl]thiourea 96%
1-[3,5-Bis(trifluoromethyl)phenyl]-3-[(1R,2R)-(-)-2-(dimethylamino)cyclohexyl]thiourea

Precaución: for Research Use Only

Formula: C₁₇H₂₁F₆N₃S
MDL Number: MFCD09834839
CAS Number: 620960-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR