2,6-Dibromo-4H-cyclopenta[1,2-b:5,4-b']dithiophen-4-one 95%

Supplier: Ambeed
Warning

A195950-100MG A195950-250MG
76825-378EA 32 USD
76825-378 76825-380
2,6-Dibromo-4H-cyclopenta[1,2-b:5,4-b']dithiophen-4-one 95%
2,6-Dibromo-4H-cyclopenta[1,2-b:5,4-b']dithiophen-4-one

Precaución: for Research Use Only

Formula: C₉H₂Br₂OS₂
MDL Number: MFCD23135467
CAS Number: 636588-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR