2-Chloro-5-cyclopropylpyridine 96%

Supplier: Ambeed
Warning

A195285-100MG A195285-1G A195285-250MG A195285-5G
76825-362EA 22 USD
76825-362 76825-364 76825-366 76825-368
2-Chloro-5-cyclopropylpyridine 96%
2-Chloro-5-cyclopropylpyridine

Precaución: for Research Use Only

Formula: C₈H₈ClN
MDL Number: MFCD12963717
CAS Number: 1042986-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR