5-Aminopyrazine-2-carboxamide 98%

Supplier: Ambeed
Warning

A194451-100MG A194451-1G A194451-250MG
76825-332EA 126.25 USD
76825-332 76825-334 76825-336
5-Aminopyrazine-2-carboxamide 98%
5-Aminopyrazine-2-carboxamide

Precaución: for Research Use Only

Formula: C₅H₆N₄O
MW: 138.13 g/mol
MDL Number: MFCD08705774
CAS Number: 89323-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR