3-Methyl-[1,2,4]triazolo[4,3-a]pyridine 98%

Supplier: Ambeed
Warning

A193488-1G A193488-250MG A193488-5G
76825-324EA 136.86 USD
76825-324 76825-326 76825-328
3-Methyl-[1,2,4]triazolo[4,3-a]pyridine 98%
3-Methyl-[1,2,4]triazolo[4,3-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₇N₃
MW: 133.15 g/mol
MDL Number: MFCD00498952
CAS Number: 1004-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR