2-Phenyl-1H-indol-5-amine 97%

Supplier: Ambeed
Warning

A192984-250MG A192984-1G
76825-322EA 101 USD
76825-322 77885-433
2-Phenyl-1H-indol-5-amine 97%
2-Phenyl-1H-indol-5-amine

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂
MDL Number: MFCD17012057
CAS Number: 6855-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR