3-Chloro-2,4-difluoropyridine 95%

Supplier: Ambeed
Warning

A192627-100MG A192627-250MG
76825-314EA 74.27 USD
76825-314 76825-316
3-Chloro-2,4-difluoropyridine 95%
3-Chloro-2,4-difluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂ClF₂N
MW: 149.526 g/mol
MDL Number: MFCD09998888
CAS Number: 851179-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR