9-Boc-3-Oxa-9-azabicyclo[3.3.1]nonan-7-one 97%

Supplier: Ambeed
Warning

A192285-1G A192285-250MG
76825-310EA 54 USD
76825-310 76825-312
9-Boc-3-Oxa-9-azabicyclo[3.3.1]nonan-7-one 97%
9-Boc-3-Oxa-9-azabicyclo[3.3.1]nonan-7-one

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₄
MDL Number: MFCD17016705
CAS Number: 280761-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR