Methyl 3,5-dichloropicolinate 98%

Supplier: Ambeed
Warning

A192251-1G A192251-250MG
76825-306EA 318.27 USD
76825-306 76825-308
Methyl 3,5-dichloropicolinate 98%
Methyl 3,5-dichloropicolinate

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
MW: 206.02 g/mol
MDL Number: MFCD09839147
CAS Number: 5439-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR