2-Amino-N-(o-tolyl)benzamide 96%

Supplier: Ambeed
Warning

A191613-1G A191613-25G A191613-5G
76825-288EA 10 USD
76825-288 76825-290 76825-292
2-Amino-N-(o-tolyl)benzamide 96%
2-Amino-N-(o-tolyl)benzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₄N₂O
MDL Number: MFCD00578802
CAS Number: 4943-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR