5-Amino-3-methylbenzo[d]isoxazole 95%

Supplier: Ambeed
Warning

A191241-100MG A191241-1G A191241-250MG
76825-282EA 50 USD
76825-282 76825-284 76825-286
5-Amino-3-methylbenzo[d]isoxazole 95%
5-Amino-3-methylbenzo[d]isoxazole

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.16 g/mol
MDL Number: MFCD11111683
CAS Number: 851768-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR