(6-Chloropyridin-2-yl)methanamine dihydrochloride 95%

Supplier: Ambeed
Warning

A191190-1G A191190-250MG A191190-5G
76825-276EA 112 USD
76825-276 76825-278 76825-280
(6-Chloropyridin-2-yl)methanamine dihydrochloride 95%
(6-Chloropyridin-2-yl)methanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₇ClN₂·2ClH
MDL Number: MFCD11046128
CAS Number: 1557921-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR