6-Chloro-3-nitropyridine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A190677-100MG A190677-250MG
76825-272EA 32 USD
76825-272 76825-274
6-Chloro-3-nitropyridine-2-carboxylic acid 95%
6-Chloro-3-nitropyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃ClN₂O₄
MDL Number: MFCD00661262
CAS Number: 1204400-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR