4,6-Dichloropyrido[3,2-d]pyrimidine 97%

Supplier: Ambeed
Warning

A190570-100MG A190570-1G A190570-250MG A190570-5G
76825-264EA 21 USD
76825-264 76825-266 76825-268 76825-270
4,6-Dichloropyrido[3,2-d]pyrimidine 97%
4,6-Dichloropyrido[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₃Cl₂N₃
MDL Number: MFCD11846204
CAS Number: 175358-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR