2-Propoxy-3-pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Propoxypyridine-3-boronic acid, (2-Propoxypyridin-3-yl)boronic acid

A190194-100MG A190194-1G A190194-250MG
76825-246EA 36.08 USD
76825-246 76825-248 76825-250
2-Propoxy-3-pyridineboronic acid 98%
2-Propoxy-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₂BNO₃
MW: 181 g/mol
MDL Number: MFCD13195680
CAS Number: 1218790-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR