(4-Methylpiperazin-1-yl)(4-nitrophenyl)methanone 95%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-4-[(4-nitrophenyl)carbonyl]piperazine

A189921-1G A189921-250MG
76825-238EA 31.83 USD
76825-238 76825-240
(4-Methylpiperazin-1-yl)(4-nitrophenyl)methanone 95%
(4-Methylpiperazin-1-yl)(4-nitrophenyl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₅N₃O₃
MW: 249.27 g/mol
MDL Number: MFCD00585638
CAS Number: 21091-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR