2-Fluoro-5-(propylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-Fluoro-5-(propylcarbamoyl)phenyl)boronic acid

A189740-1G A189740-250MG A189740-5G
76825-226EA 123.07 USD
76825-226 76825-228 76825-230
2-Fluoro-5-(propylcarbamoyl)phenylboronic acid 98%
2-Fluoro-5-(propylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BFNO₃
MW: 225.0300 g/mol
MDL Number: MFCD08436029
CAS Number: 874289-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR