(2R,4R)-Ethyl 1-(2-amino-5-(3-nitroguanidino)pentanoyl)-4-methylpiperidine-2-carboxylate hydrochloride 95%

Supplier: Ambeed
Danger

A189669-1G A189669-250MG
76825-222EA 35.01 USD
76825-222 76825-224
(2R,4R)-Ethyl 1-(2-amino-5-(3-nitroguanidino)pentanoyl)-4-methylpiperidine-2-carboxylate hydrochloride 95%
(2R,4R)-Ethyl 1-(2-amino-5-(3-nitroguanidino)pentanoyl)-4-methylpiperidine-2-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₂₉ClN₆O₅
MW: 408.88 g/mol
MDL Number: MFCD11982553
CAS Number: 74874-08-1
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR