6-Nitrooxindole 95%

Supplier: Ambeed
Warning

Synonyms: 6-Nitroindolin-2-one

A189570-100MG A189570-250MG
76825-218EA 28.65 USD
76825-218 76825-220
6-Nitrooxindole 95%
6-Nitrooxindole

Precaución: for Research Use Only

Formula: C₈H₆N₂O₃
MW: 178.145 g/mol
MDL Number: MFCD09835636
CAS Number: 474799-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR