2-Chloro-4-formylphenylboronic acid 98%

Supplier: Ambeed
Warning

A189439-100MG A189439-1G A189439-250MG A189439-25G A189439-5G
76825-208EA 23.34 USD
76825-208 76825-210 76825-212 76825-214 76825-216
2-Chloro-4-formylphenylboronic acid 98%
2-Chloro-4-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BClO₃
MW: 184.385 g/mol
MDL Number: MFCD18311828
CAS Number: 1063712-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR