2-Fluoro-5-methoxy-4-nitrobenzoic acid 98%

Supplier: Ambeed
Danger

A189107-100MG A189107-1G A189107-250MG A189107-5G
76825-196EA 10 USD
76825-196 76825-198 76825-200 76825-202
2-Fluoro-5-methoxy-4-nitrobenzoic acid 98%
2-Fluoro-5-methoxy-4-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆FNO₅
MDL Number: MFCD26395470
CAS Number: 1001345-80-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR