5-Acetyl-1-methyl-1H-indazole 98%

Supplier: Ambeed
Warning

Synonyms: 5-Ethanoyl-1-methyl-1H-indazole, 1-(1-Methyl-1H-indazol-5-yl)ethan-1-one, 1-(1-Methyl-1H-indazol-5-yl)ethanone

A188580-100MG A188580-1G A188580-250MG
76825-174EA 68.96 USD
76825-174 76825-176 76825-178
5-Acetyl-1-methyl-1H-indazole 98%
5-Acetyl-1-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O
MW: 174.199 g/mol
MDL Number: MFCD11869767
CAS Number: 1159511-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR