Iridium(IV) chloride 95%

Supplier: Ambeed
Warning

A188331-100MG A188331-1G A188331-250MG
76825-162EA 58.35 USD
76825-162 76825-164 76825-166
Iridium(IV) chloride 95%
Iridium(IV) chloride

Precaución: for Research Use Only

Formula: Cl₄Ir
MW: 334.03 g/mol
MDL Number: MFCD00016155
CAS Number: 10025-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR