1,3,5-Tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene 97%

Supplier: Ambeed
Warning

Synonyms: 2,2',2''-(1,3,5-Benzenetriyl)tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolane), 1,3,5-Phenyltriboronic acid tris(pinacol) ester

A188038-100G A188038-10G A188038-1G A188038-250MG A188038-25G A188038-5G
76825-144EA 994.07 USD
76825-144 76825-146 76825-148 76825-150 76825-152 76825-154
1,3,5-Tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene 97%
1,3,5-Tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

Precaución: for Research Use Only

Formula: C₂₄H₃₉B₃O₆
MW: 456.00 g/mol
MDL Number: MFCD17167305
CAS Number: 365564-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR