SB-525334 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-[2-(tert-Butyl)-5-(6-methylpyridin-2-yl)-1H-imidazol-4-yl]quinoxaline, 6-(2-(tert-Butyl)-5-(6-methylpyridin-2-yl)-1H-imidazol-4-yl)quinoxaline
A187949-10MG
A187949-1MG
A187949-50MG
A187949-5MG
A187949-100MG
76825-134EA
59.42
USD
76825-134
76825-136
76825-138
76825-140
77683-481
SB-525334 98+%
SB-525334
Precaución: For research use only.
Formula:
C₂₁H₂₁N₅ MW: 343.43 g/mol |
MDL Number:
MFCD11045307 CAS Number: 356559-20-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...