1-tert-Butyl 4-ethyl 4-cyanopiperidine-1,4-dicarboxylate 95%

Supplier: Ambeed
Warning

A186446-100MG A186446-1G A186446-250MG
76825-106EA 67.9 USD
76825-106 76825-108 76825-110
1-tert-Butyl 4-ethyl 4-cyanopiperidine-1,4-dicarboxylate 95%
1-tert-Butyl 4-ethyl 4-cyanopiperidine-1,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₂N₂O₄
MW: 282.34 g/mol
MDL Number: MFCD15144556
CAS Number: 1016258-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR