2-Amino-5-iodo-4-(trifluoromethyl)-1,3-thiazole 95%

Supplier: Ambeed
Warning

Synonyms: 5-Iodo-4-(trifluoromethyl)-1,3-thiazol-2-amine, 5-Iodo-4-(trifluoromethyl)thiazol-2-amine

A186125-100MG A186125-1G A186125-250MG A186125-5G
76825-094EA 138.98 USD
76825-094 76825-096 76825-098 76825-100
2-Amino-5-iodo-4-(trifluoromethyl)-1,3-thiazole 95%
2-Amino-5-iodo-4-(trifluoromethyl)-1,3-thiazole

Precaución: for Research Use Only

Formula: C₄H₂F₃IN₂S
MW: 294.037 g/mol
MDL Number: MFCD11847617
CAS Number: 682342-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR