Boc-Lysinol(Z) 96%

Supplier: Ambeed
Warning

Synonyms: Benzyl tert-butyl (6-hydroxyhexane-1,5-diyl)(S)-dicarbamate, Boc-Lys(Z)-ol

A185875-10G A185875-1G A185875-5G
76825-088EA 138.98 USD
76825-088 76825-090 77593-196
Boc-Lysinol(Z) 96%
Boc-Lysinol(Z)

Precaución: for Research Use Only

Formula: C₁₉H₃₀N₂O₅
MW: 366.5 g/mol
MDL Number: MFCD00235934
CAS Number: 82689-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR