4-(Trifluoromethyl)piperidin-4-ol 98%

Supplier: Ambeed
Warning

A185307-1G A185307-5G A185307-100MG A185307-250MG
76825-072EA 94.43 USD
76825-072 76825-074 77683-468 77683-469
4-(Trifluoromethyl)piperidin-4-ol 98%
4-(Trifluoromethyl)piperidin-4-ol

Precaución: For research use only.

Formula: C₆H₁₀F₃NO
MW: 169.145 g/mol
MDL Number: MFCD13179174
CAS Number: 373603-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR