3,5-Dibromo-1-methyl-1H-1,2,4-triazole 98%

Supplier: Ambeed
Warning

A185259-1G A185259-250MG A185259-25G A185259-5G
76825-064EA 84.88 USD
76825-064 76825-066 76825-068 76825-070
3,5-Dibromo-1-methyl-1H-1,2,4-triazole 98%
3,5-Dibromo-1-methyl-1H-1,2,4-triazole

Precaución: for Research Use Only

Formula: C₃H₃Br₂N₃
MDL Number: MFCD09426402
CAS Number: 23579-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR