Methyl 5-bromo-2-hydroxy-4-methylbenzoate 97%

Supplier: Ambeed
Warning

A184895-1G A184895-250MG A184895-25G
76825-058EA 18 USD
76825-058 76825-060 77593-188
Methyl 5-bromo-2-hydroxy-4-methylbenzoate 97%
Methyl 5-bromo-2-hydroxy-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MDL Number: MFCD11045602
CAS Number: 39503-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR