Methyl 5-bromo-6-methoxypicolinate 97%

Supplier: Ambeed
Warning

A184209-10G A184209-250MG A184209-5G
76825-040EA 758.55 USD
76825-040 76825-042 76825-044
Methyl 5-bromo-6-methoxypicolinate 97%
Methyl 5-bromo-6-methoxypicolinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
MW: 246.05 g/mol
MDL Number: MFCD14698129
CAS Number: 1214329-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR