Bis(4-(2,4,4-trimethylpentan-2-yl)phenyl)amine 97%
Supplier: Ambeed
|
Warning
|
A183779-10G
A183779-1G
A183779-250MG
A183779-25G
A183779-5G
76825-010EA
121
USD
76825-010
76825-012
76825-014
76825-016
76825-018
Bis(4-(2,4,4-trimethylpentan-2-yl)phenyl)amine 97%
Bis(4-(2,4,4-trimethylpentan-2-yl)phenyl)amine
Precaución: for Research Use Only
Formula:
C₂₈H₄₃N |
MDL Number:
MFCD00345316 CAS Number: 15721-78-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...