Methyl 5-bromo-2-methylnicotinate 98%

Supplier: Ambeed
Warning

A183772-1G A183772-250MG A183772-5G
76825-004EA 92.3 USD
76825-004 76825-006 76825-008
Methyl 5-bromo-2-methylnicotinate 98%
Methyl 5-bromo-2-methylnicotinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.05 g/mol
MDL Number: MFCD13705366
CAS Number: 1215916-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR