4-Chloro-3-(trifluoromethyl)benzamide 96%

Supplier: Ambeed
Warning

A183700-10G A183700-1G A183700-250MG
76824-998EA 50.93 USD
76824-998 76825-000 76825-002
4-Chloro-3-(trifluoromethyl)benzamide 96%
4-Chloro-3-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₈H₅ClF₃NO
MW: 223.58 g/mol
MDL Number: MFCD04115832
CAS Number: 62584-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR