(3-(2-Hydroxypropan-2-yl)phenyl)boronic acid 97%
Supplier: Ambeed
|
Warning
|
A183694-100MG
A183694-1G
A183694-250MG
A183694-5G
76824-992EA
38.2
USD
76824-992
76824-994
76824-996
77683-462
(3-(2-Hydroxypropan-2-yl)phenyl)boronic acid 97%
(3-(2-Hydroxypropan-2-yl)phenyl)boronic acid
Precaución: For research use only.
Formula:
C₉H₁₃BO₃ MW: 180.009 g/mol |
MDL Number:
MFCD09992906 CAS Number: 955369-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...