2-Bromo-3,4,5-trifluoroaniline 95%

Supplier: Ambeed
Warning

A183535-1G A183535-250MG A183535-25G A183535-5G
76824-984EA 37.14 USD
76824-984 76824-986 76824-988 76824-990
2-Bromo-3,4,5-trifluoroaniline 95%
2-Bromo-3,4,5-trifluoroaniline

Precaución: for Research Use Only

Formula: C₆H₃BrF₃N
MW: 225.9939 g/mol
MDL Number: MFCD01311010
CAS Number: 1000577-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR