2-(2,5-Dibromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dibromophenylboronic acid pinacol ester

A183449-100MG A183449-1G A183449-250MG A183449-5G
76824-976EA 9.55 USD
76824-976 76824-978 76824-980 76824-982
2-(2,5-Dibromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-(2,5-Dibromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₂H₁₅BBr₂O₂
MW: 361.865 g/mol
MDL Number: MFCD17215817
CAS Number: 1256781-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR