(S)-1-(3,5-Dichloropyridin-4-yl)ethanol 97%

Supplier: Ambeed
Warning

A182828-100MG A182828-1G A182828-250MG A182828-5G
76824-966EA 16 USD
76824-966 76824-968 76824-970 76824-972
(S)-1-(3,5-Dichloropyridin-4-yl)ethanol 97%
(S)-1-(3,5-Dichloropyridin-4-yl)ethanol

Precaución: for Research Use Only

Formula: C₇H₇Cl₂NO
MDL Number: MFCD22665797
CAS Number: 1370347-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR