(-)-Monomenthyl succinate 95%

Supplier: Ambeed
Warning

Synonyms: Succinic acid (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl ester, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl succinate, (-)-Menthyl succinate, Succinic acid (-)-menthyl Ester, 4-(((1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl)oxy)-4-oxobutanoic acid

A181838-100G A181838-10G A181838-5G
76824-952EA 174 USD
76824-952 76824-954 76824-956
(-)-Monomenthyl succinate 95%
(-)-Monomenthyl succinate

Precaución: for Research Use Only

Formula: C₁₄H₂₄O₄
MDL Number: MFCD00002788
CAS Number: 77341-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR