1-Cyclopropyl-4-iodo-1H-pyrazole 96%

Supplier: Ambeed
Warning

A181829-1G A181829-250MG A181829-5G
76824-948EA 129.43 USD
76824-948 76824-950 77683-458
1-Cyclopropyl-4-iodo-1H-pyrazole 96%
1-Cyclopropyl-4-iodo-1H-pyrazole

Precaución: For research use only.

Formula: C₆H₇IN₂
MDL Number: MFCD22573364
CAS Number: 1239363-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR