2,3-Dimethyl-2H-indazol-6-amine hydrochloride 97%

Supplier: Ambeed
Warning

A181727-10G A181727-1G A181727-25G A181727-5G
76824-936EA 48.81 USD
76824-936 76824-938 76824-940 76824-942
2,3-Dimethyl-2H-indazol-6-amine hydrochloride 97%
2,3-Dimethyl-2H-indazol-6-amine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClN₃
MW: 197.67 g/mol
MDL Number: MFCD09258896
CAS Number: 635702-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR