3-Chloro-2-fluoro-4-pyridineboronic acid 97%

Supplier: Ambeed
Warning

A181076-1G A181076-100MG A181076-250MG
76824-920EA 38 USD
76824-920 77885-396 77885-397
3-Chloro-2-fluoro-4-pyridineboronic acid 97%
3-Chloro-2-fluoro-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₄BClFNO₂
MDL Number: MFCD09037477
CAS Number: 1217500-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR