1-(2,4-Dihydroxy-5-isopropylphenyl)ethanone 97%

Supplier: Ambeed
Warning

A181055-100MG A181055-1G A181055-5G
76824-914EA 31.83 USD
76824-914 76824-916 76824-918
1-(2,4-Dihydroxy-5-isopropylphenyl)ethanone 97%
1-(2,4-Dihydroxy-5-isopropylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
MDL Number: MFCD12827976
CAS Number: 747414-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR