2-Bromo-3-methylanisole 95%

Supplier: Ambeed
Warning

A180897-10G A180897-1G A180897-25G A180897-5G A180897-250MG
76824-906EA 224 USD
76824-906 76824-908 76824-910 76824-912 77885-394
2-Bromo-3-methylanisole 95%
2-Bromo-3-methylanisole

Precaución: for Research Use Only

Formula: C₈H₉BrO
MDL Number: MFCD14705101
CAS Number: 38197-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR