Methyl 4-iodo-1-(4-methoxybenzyl)-1H-pyrazole-3-carboxylate 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A180828-100MG A180828-1G A180828-5G A180828-250MG
76824-898EA 60 USD
76824-898 77683-450 77683-451 77885-393
Methyl 4-iodo-1-(4-methoxybenzyl)-1H-pyrazole-3-carboxylate 97%, Ambeed.Inc
Methyl 4-iodo-1-(4-methoxybenzyl)-1H-pyrazole-3-carboxylate

Precaución: For research use only.

Formula: C₁₃H₁₃IN₂O₃
MDL Number: MFCD15071116
CAS Number: 1260656-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR